missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Class B Clear Glass Volumetric Flask

Brand:  Duran™ 246707356

Code : NEW

Additional Details : Weight : 1.28800kg

 View more versions of this product

Product Code. 15280926

  • £133.43 / Each


Estimated Shipment
19-02-2018
Add to basket

Specifications

Specifications

ISO 1042, USP
Polyethylene
Clear
DURAN™
475mm
Class B
NS 34/35
5000mL
Yes
1Pack
Videos