missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Class B Clear Glass Volumetric Flask

Brand:  Duran™ 246706351

Code : NEW

Additional Details : Weight : 0.92000kg

 View more versions of this product

Product Code. 15270926

  • £160.00 / Pack of 2


Estimated Shipment
16-05-2018
Add to basket

Specifications

Specifications

ISO 1042, USP
Polyethylene
Clear
DURAN™
370mm
Class B
NS 29/32
2000mL
Yes
2Pack
Videos