missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Class B Clear Glass Volumetric Flask

Brand:  Duran™ 246704453

Code : NEW

Additional Details : Weight : 0.33000kg

 View more versions of this product

Product Code. 15250926

  • £67.85 / Pack of 2


Estimated Shipment
13-02-2018
Add to basket

Specifications

Specifications

ISO 1042, USP
Polyethylene
Clear
DURAN™
260mm
Class B
NS 19/26
500mL
Yes
2Pack
Videos