missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann Instruments™ Class A Measuring Cylinders, Blue Graduation

Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann Instruments™ Class A Measuring Cylinders, Blue Graduation are manufactured from durable DURAN™ glass tubing.

Brand:  Hirschmann Instruments™ 224017027

Code : A0

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 10747793

  • £19.00 / Pack of 2


Estimated Shipment
21-06-2018
Add to basket

Description

Description

  • With spout, main point ring graduation
  • Blue graduation
  • Conformity certified, with dated batch identification
  • Acc. to DIN EN ISO 4788
  • Corrosion-free and resistant to thermal shock
Specifications

Specifications

170mm
A
0.25
0.25
DIN EN ISO 4788
25mL
0.5mL
Videos