missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann Instruments™ Class A Measuring Cylinders, Blue Graduation, with Plastic Socket

Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann Instruments™ Class A Measuring Cylinders, Blue Graduation, with Plastic Socket are manufactured from durable DURAN™ glass tubing.

Brand:  Hirschmann Instruments™ 2270170

Code : A0

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 11766692

  • £14.75 / Pack of 2


Estimated Shipment
25-06-2018
Add to basket

Description

Description

  • With spout and plastic socket, main point ring graduation
  • Blue graduation
  • Conformity certified, with dated batch information
  • Acc. to DIN EN ISO 4788
  • Corrosion-free and resistant to thermal shock
Specifications

Specifications

170mm
DIN EN ISO 4788
25mL
0.5mL
Glass (Untyped)
A
0.25
0.25
Videos
View accessory products