missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann Instruments™ Class A Measuring Cylinders, Blue Graduation, with Plastic Socket

Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann Instruments™ Class A Measuring Cylinders, Blue Graduation, with Plastic Socket are manufactured from durable DURAN™ glass tubing.

Brand:  Hirschmann Instruments™ 2270160

Code : A0

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 11756692

  • £14.70 / Pack of 2


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • With spout and plastic socket, main point ring graduation
  • Blue graduation
  • Conformity certified, with dated batch information
  • Acc. to DIN EN ISO 4788
  • Corrosion-free and resistant to thermal shock
Specifications

Specifications

140mm
DIN EN ISO 4788
10mL
0.2mL
Glass (Untyped)
A
0.1
0.1
Videos