missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Class A Measuring Cylinders

Features a hexagonal base. DURAN™ Class A Measuring Cylinders blue gradations make them easy to read.

Brand:  Duran™ 213901406

Code : NEW

Additional Details : Weight : 0.11000kg

 View more versions of this product

Product Code. 15260846

  • £22.85 / Pack of 2


Estimated Shipment
19-02-2018
Add to basket

Description

Description

  • Blue graduation, with main points ring graduation and hexagonal base
  • Certificate of conformity "H"
Specifications

Specifications

167mm
A
Blue graduation
0.5mL
ISO 4788
25mL
±0.25mL
121°C
Videos