missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Class A Clear Glass Volumetric Flask

Brand:  Duran™ 246716352

Code : NEW

Additional Details : Weight : 0.92000kg

 View more versions of this product

Product Code. 15280936

  • £250.00 / Pack of 2


Estimated Shipment
11-05-2018
Add to basket

Specifications

Specifications

ISO 1042, USP and H
Polyethylene
Clear
DURAN™
370mm
Class A
NS 29/32
2000mL
Yes
2Pack
Videos