missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Class A Clear Glass Volumetric Flask

Brand:  Duran™ 246714454

Code : NEW

Additional Details : Weight : 0.33000kg

 View more versions of this product

Product Code. 15260936

  • £143.00 / Pack of 2


Estimated Shipment
13-02-2018
Add to basket

Specifications

Specifications

ISO 1042, USP and H
Polyethylene
Clear
DURAN™
260mm
Class A
NS 19/26
500mL
Yes
2Pack
Videos