missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Class A Clear Glass Volumetric Flask

Brand:  Duran™ 246713655

Code : NEW

Additional Details : Weight : 0.18000kg

 View more versions of this product

Product Code. 15250936

  • £131.00 / Pack of 2


Estimated Shipment
20-06-2018
Add to basket

Specifications

Specifications

ISO 1042, USP and H
Polyethylene
Clear
Borosilicate GlassþDURAN™
220mm
Class A
NS 14/23
250mL
Yes
2Pack
Videos