missing translation for 'onlineSavingsMsg'
Learn More

Poulten Graf™ Class A Amber Borosilicate Glass Trapezoidal Volumetric Flask

Created from borosilicate glass. Poulten Graf™ Volumetric Flasks with Class A Stopper are used to prepare standard solutions.

Brand:  Poulten Graf™ RB257/AM/A/5

Code : 01

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12373976

  • £99.95 / Pack of 5


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Polypropylene stopper included
  • Each flask individually numbered for identification and traceability
  • Volac™ borosilicate glass
Specifications

Specifications

Trapezoidal
Polypropylene
Amber
Borosilicate Glass
5
Class A
7/16
1mL
Yes
±0.010mL
Videos
Documentation