missing translation for 'onlineSavingsMsg'
Learn More

Poulten Graf™ Class A Borosilicate Glass Volumetric Flask without Certificate of Calibration

Brand:  Poulten Graf™ US258/E/5

Code : 01

Additional Details : Weight : 0.01000kg

Product Code. 12383896

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

Single mark graduation and interchangeable polypropylene stopper. Each flask has been individually tested and carries its own individual number for full identification and traceability.
Specifications

Specifications

USP, ISO 1042, DIN 12664
Red
10/19
25mL
Yes
0.03mL
Class A
Polyethylene
Clear
Borosilicate Glass
5
Videos
Documentation