missing translation for 'onlineSavingsMsg'
Learn More

Poulten Graf™ Class A Borosilicate Glass Volumetric Flask with Certificate of Calibration

Allows for precise and safe liquid handling in the laboratory. Poulten Graf™ Class A Borosilicate Glass Volumetric Flasks with Certificate of Calibration are individually printed, numbered, and supplied with an individual correction certificate of accuracy.

Brand:  Poulten Graf™ US258/AM/WAC/L/S

Code : 01

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12343946

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Allows for precise and safe liquid handling in the laboratory. Poulten Graf™ Class A Borosilicate Glass Volumetric Flasks with Certificate of Calibration are individually printed, numbered, and supplied with an individual correction certificate of accuracy.

  • Tooled and ground neck fitted with a Polystop™ polypropylene interchangeable stopper
  • Single mark gradation
  • Available in amber or clear glass
  • Each flask carries an individual number for identification and traceability
Specifications

Specifications

USP, ISO 1042, DIN 12664
Polyethylene
2000 mL
1
Yes
10
Red
Amber
Borosilicate Glass
0.50 mL
A
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers