missing translation for 'onlineSavingsMsg'
Learn More

Poulten Graf™ Class A Borosilicate Glass Volumetric Flask with Certificate of Calibration

Allows for precise and safe liquid handling in the laboratory. Poulten Graf™ Class A Borosilicate Glass Volumetric Flasks with Certificate of Calibration are individually printed, numbered, and supplied with an individual correction certificate of accuracy.

Brand:  Poulten Graf™ US258/WAC/F/5

Code : 01

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 10521563

  • £127.00 / Pack of 5


Estimated Shipment
16-07-2018
Add to basket

Description

Description

  • Tooled and ground neck fitted with a Polystop™ polypropylene interchangeable stopper
  • Single mark gradation
  • Available in amber or clear glass
  • Each flask carries an individual number for identification and traceability
Specifications

Specifications

USP, ISO 1042, DIN 12664
Polypropylene
Clear
Borosilicate Glass
5
Class A
10
50mL
Yes
0.05mL
Videos
Documentation