missing translation for 'onlineSavingsMsg'
Learn More

Poulten Graf™ Class A Borosilicate Glass Volumetric Flask with Certificate of Calibration

Allows for precise and safe liquid handling in the laboratory. Poulten Graf™ Class A Borosilicate Glass Volumetric Flasks with Certificate of Calibration are individually printed, numbered, and supplied with an individual correction certificate of accuracy.

Brand:  Poulten Graf™ US258/AM/WAC/F/5

Code : 01

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10529482

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Allows for precise and safe liquid handling in the laboratory. Poulten Graf™ Class A Borosilicate Glass Volumetric Flasks with Certificate of Calibration are individually printed, numbered, and supplied with an individual correction certificate of accuracy.

  • Tooled and ground neck fitted with a Polystop™ polypropylene interchangeable stopper
  • Single mark gradation
  • Available in amber or clear glass
  • Each flask carries an individual number for identification and traceability
Specifications

Specifications

USP, ISO 1042, DIN 12664
Polyethylene
50 mL
5
Yes
12.5/21
Red
Amber
Borosilicate Glass
0.05 mL
A
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers