missing translation for 'onlineSavingsMsg'
Learn More

Poulten Graf™ Class A Borosilicate Glass Volumetric Flask with Certificate of Calibration

Allows for precise and safe liquid handling in the laboratory. Poulten Graf™ Class A Borosilicate Glass Volumetric Flasks with Certificate of Calibration are individually printed, numbered, and supplied with an individual correction certificate of accuracy.

Brand:  Poulten Graf™ US258/AM/WAC/J/2

UNSPSC : 41121804

Code : 01

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10727534

  • £157.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

  • Tooled and ground neck fitted with a Polystop™ polypropylene interchangeable stopper
  • Single mark gradation
  • Available in amber or clear glass
  • Each flask carries an individual number for identification and traceability
Specifications

Specifications

USP, ISO 1042, DIN 12664
Red
18.8/26
Yes
0.15mL
Borosilicate Glass
Class A
Polyethylene
Amber
2
500mL
Documentation