Learn More
Poulten & Graf GmbH Volumetric Flask with Class A Stopper
Created from borosilicate glass. Poulten Graf™ FORTUNA™ Borosilicate Glass Class A Volumetric Flasks offers excellent chemical resistance and are best for obtaining a precise measurement of chemical solutions.
Brand: Poulten & Graf GmbH RB258/AM/A/5
Code : 01
Additional Details : Weight : 0.15792kg
Description
With class A stopper
Amber color
Manufactured from borosilicate glass
Specifications
ISO and ASTM certified | |
5 mL | |
5 | |
A | |
10/19 |
Amber | |
Borosilicate Glass | |
Yes | |
Polystop Stopper |