missing translation for 'onlineSavingsMsg'
Learn More

Poulten Graf™ Class A Amber Borosilicate Glass Trapezoidal Volumetric Flask

Shaped to maximize stability while allowing access to contents. Poulten Graf™ Amber Borosilicate Glass Trapezoidal Volumetric Flasks have tight accuracy tolerances and are individually calibrated and tested.

Brand:  Poulten Graf™ RB257/C/5

Code : 01

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12343976

  • £76.60 / Pack of 5


In Stock 8
Add to basket

Description

Description

  • Polypropylene stopper included
  • Each flask individually numbered for identification and traceability
  • Volac™ borosilicate glass
Specifications

Specifications

Trapezoidal
Polypropylene
Clear
Borosilicate Glass
5
Class A
7/16
3mL
Yes
±0.025mL
Videos
Documentation