missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Class A Amber Glass Volumetric Flask with Individual Certificate

Brand:  Duran™ 246773652

Code : NEW

Additional Details : Weight : 0.18000kg

 View more versions of this product

Product Code. 15230916

  • £166.00 / Pack of 2


Estimated Shipment
16-03-2018
Add to basket

Specifications

Specifications

ISO 1042 and H
Polyethylene
Amber
DURAN™
220mm
Class A
NS 14/23
250mL
Yes
2Pack
Videos