missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Class A Amber Glass Volumetric Flask with Individual Certificate

Brand:  Duran™ 246775456

Code : NEW

Additional Details : Weight : 0.49000kg

 View more versions of this product

Product Code. 15250916

  • £266.00 / Pack of 2


Estimated Shipment
13-06-2018
Add to basket

Specifications

Specifications

ISO 1042 and H
Polyethylene
Amber
DURAN™
300mm
Class A
NS 24/29
1000mL
Yes
2Pack
Videos