Learn More
Thermo Scientific™ Clamps for CO2 Resistant Shaker
Achieve maximize capacity on the Thermo Scientific™ CO2 Resistant Shaker universal platform with a variety of clamps to fit most vessels.
Brand: Thermo Scientific™ 88881105
Code : NEW
Additional Details : Weight : 0.08930kg
Description
Maximize capacity on the Thermo Scientific™ CO2 Resistant Shaker universal platform with a variety of clamps to fit most vessels.
Specifications
Flask Clamp | |
50mL clamp | |
CO2 Resistant Shaker | |
30 |