missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CL-XPosure™ Film (X-Ray Film)

Capture results of ECL-based Western blots and other chemiluminescent protein or nucleic acid assays using this economically priced, clear-blue X-ray film.

Brand:  Thermo Scientific™ 34097

Code : PN

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10571204

  • £228.93 / 100 sheets


Estimated Shipment
15-03-2018
Add to basket

Description

Description

CL-XPosure Film is an excellent phototgraphic film for use with enhanced chemiluminescence (ECL) substrates for horseradish peroxidase (HRP) or alkaline phosphatase (AP). Convenient sizes are offered for protein Western blotting, standard film cassettes and film developing equipment. The autoradiography film also works well for chemiluminescent or isotopic DNA and RNA methods, including Southern and Northern blotting and gel-shift assays (EMSA). The quality and price of this film make it the smart choice for life science research applications.

Highlights:

  • High performance – just as sensitive as other commercially available autoradiography films

  • Affordable – costs up to 75% less than other brands of comparable quality X-ray film

  • Convenient – available in five sheet sizes in packages containing 50 or 100 non-interleaved sheets

  • Perfect match – validated for use with Thermo Scientific SuperSignal Chemiluminescent Substrates and Thermo Scientific Pierce ECL Chemiluminescent Substrate

Ordering Information:

Available in 5 × 7in., 7 × 9.5in. , 9.5 × 11in., or 14 × 17in. sheets

Specifications

Specifications

CL-XPosure Film, 9.5 x 11.8in. (24 x 30cm)
Blue-backed, silver-emulsion, two-sided autoradiography film
100 sheets
100 uses
Videos
Documentation

Technical Documents