missing translation for 'onlineSavingsMsg'
Learn More

Cl-4AS-1, Tocris Bioscience™

Steroidal androgen receptor agonist

Brand:  Tocris (Bio-Techne) 381250

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13463634

  • £728.28 / 50mg


Estimated Shipment
29-10-2018
Add to basket

Description

Description

Potent steroidal androgen receptor agonist (IC50 = 12 nM). Mimics the action of 5α-dihydrotestosterone (DHT). Transactivates the mouse mammary tumor virus (MMTV) promoter; represses MMP1 promoter activity. Inhibits 5α-reductase type I and II (IC50 values are 6 and 10 nM respectively).
Specifications

Chemical Identifiers

188589-66-4
441.012
cl-4as-1, 4a?,4b?,6a?,7?,9a?,9b?,11a?-n-2-chlorophenyl-2,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-tetradecahydro-1,4,6a-trimethyl-2-oxo-1h-indeno 5,4-f quinoline-7-carboxamide
(1S,3aS,3bS,9aR,9bS,11aR)-N-(2-chlorophenyl)-6,9a,11a-trimethyl-7-oxo-2,3,3a,3b,4,5,5a,9b,10,11-decahydro-1H-indeno[5,4-f]quinoline-1-carboxamide
C26H33ClN2O2
CTVXDPDUOKQBKZ-BLIBDJTASA-N
90488881
CC12CCC3C(C1CCC2C(=O)NC4=CC=CC=C4Cl)CCC5C3(C=CC(=O)N5C)C

Specifications

Cl-4AS-1
188589-66-4
cl-4as-1, 4a?,4b?,6a?,7?,9a?,9b?,11a?-n-2-chlorophenyl-2,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-tetradecahydro-1,4,6a-trimethyl-2-oxo-1h-indeno 5,4-f quinoline-7-carboxamide
CC12CCC3C(C1CCC2C(=O)NC4=CC=CC=C4Cl)CCC5C3(C=CC(=O)N5C)C
441.012
441.01
50mg
C26H33ClN2O2
CTVXDPDUOKQBKZ-BLIBDJTASA-N
(1S,3aS,3bS,9aR,9bS,11aR)-N-(2-chlorophenyl)-6,9a,11a-trimethyl-7-oxo-2,3,3a,3b,4,5,5a,9b,10,11-decahydro-1H-indeno[5,4-f]quinoline-1-carboxamide
90488881
>99%
Videos