missing translation for 'onlineSavingsMsg'
Learn More

Citronellal Pract., 93%, ACROS Organics™

Brand:  Acros Organics 405291000

Code : 29

Additional Details : CAS Number : 106-23-0 Weight : 0.15000kg


Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10637181

  • £21.40 / 100g


EU Stock 6
Add to basket


Specifications

Chemical Identifiers

Citronellal
106-23-0
93%
(CH3)2CCHCH2CH2CH(CH3)CH2CHO
MFCD00038090
pract., 93%
≥91%
C10H18O
154.25
3, 7-Dimethyl-6-octenal

Specifications

Citronellal
106-23-0
Liquid
93%
(CH3)2CCHCH2CH2CH(CH3)CH2CHO
MFCD00038090
Glass bottle
3, 7-Dimethyl-6-octenal
Clear light yellow
pract., 93%
Authentic
≥91%
C10H18O
154.25
15, 2328
1.4440 to 1.4500 (20°C, 589nm)
0.85 to 0.87g/mL (20°C)
100g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Citronellal

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS