Learn More
Citric acid, trisodium salt dihydrate, 99%, for biochemistry, ACROS Organics™
Brand: Acros Organics 447290010
Code : A0
Additional Details : CAS Number : 6132-04-3 Weight : 1.00000kg
Packaging | Plastic bottle |
---|---|
Quantity | 1kg |
Chemical Identifiers
6132-04-3 | |
294.10 | |
trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate | |
CHEBI:32142 | |
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+] |
C6H9Na3O9 | |
NLJMYIDDQXHKNR-UHFFFAOYSA-K | |
71474 | |
trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate |
Specifications
(0.1M, water), (0.1M, water) at 0nm, 0.01 max. at 260nm, 0.01 max. at 280nm | |
6132-04-3 | |
C6H9Na3O9 | |
trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate | |
NLJMYIDDQXHKNR-UHFFFAOYSA-K | |
trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate | |
71474 | |
294.09 | |
Biochemical | |
Solubility in water: 720g/L (25°C). Other solubilities: insoluble in alcohol | |
1kg |
Citric acid, trisodium salt dihydrate | |
99% | |
NaO2CCH2C(OH)(CO2Na)CH2CO2Na·2H2O | |
Solubility in water: 720g/L (25°C). Other solubilities: insoluble in alcohol | |
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+] | |
294.10 | |
CHEBI:32142 | |
99% | |
Plastic bottle | |
>300.0°C |