missing translation for 'onlineSavingsMsg'
Learn More

Citric acid, monosodium salt, 99%, for analysis, anhydrous, Thermo Scientific Chemicals

Catalog Number p-4813416
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
18996-35-5
C6H7NaO7
214.11
SLWOXBQHKZPUNY-UHFFFAOYSA-N
bicitra, pneucid, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, sodium salt, sodium dihydrogen citrate anhydrous, citric acid, sodium salt, citric acid sodium, sodium 2-hydroxy-1,2,3-propanetricarboxylate, 2-hydroxypropane-1,2,3-tricarboxylic acid; sodium, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, sodium salt 1:?
131675399
2-hydroxypropane-1,2,3-tricarboxylic acid;molecular hydrogen;sodium
[HH].C(C(=O)O)C(CC(=O)O)(C(=O)O)O.[Na]
This item is not returnable. View return policy
18996-35-5
C6H7NaO7
214.11
SLWOXBQHKZPUNY-UHFFFAOYSA-N
bicitra, pneucid, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, sodium salt, sodium dihydrogen citrate anhydrous, citric acid, sodium salt, citric acid sodium, sodium 2-hydroxy-1,2,3-propanetricarboxylate, 2-hydroxypropane-1,2,3-tricarboxylic acid; sodium, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, sodium salt 1:?
131675399
2-hydroxypropane-1,2,3-tricarboxylic acid;molecular hydrogen;sodium
[HH].C(C(=O)O)C(CC(=O)O)(C(=O)O)O.[Na]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY
Melting Point 209.0°C to 215.0°C
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula HO2CCH2C(OH)(CO2H)CH2CO2Na
Quantity 250 g
Solubility Information Clear colorless
Formula Weight 214.11
Percent Purity 99%
Grade Analysis
Chemical Name or Material Citric acid, monosodium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'