missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric Acid Monohydrate, Certified AR for Analysis, meets analytical specification of Ph.Eur., USP, Fisher Chemical™
61.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
74.48 GBP valid until 2024-03-30
Use promo code "21663" to get your promotional price.
74.48 GBP valid until 2024-03-30
Use promo code "21741" to get your promotional price.
Use promo code "22914" to get your promotional price.
74.48 GBP valid until 2024-03-30
Use promo code "21663" to get your promotional price.
74.48 GBP valid until 2024-03-30
Use promo code "21741" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Description
C6H8O7.H2O, CAS Number-5949-29-1, 5kg, White, 210.15g/mol, 135 deg.C, 149972, Plastic keg, 1.8, Solid
Specifications
pH | 1.8 |
Melting Point | 135°C |
Color | White |
Formula Weight | 210.15g/mol |
Quantity | 1 kg |
Packaging | Plastic powder jar |
Physical Form | Solid |
Safety and Handling
Product Identifier
- Citric acid monohydrate
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Citric acid monohydrate, Citric acid
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'