missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Citric acid monohydrate, 99+%, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 135.0°C to 152.0°C |
Flash Point | 174°C |
Ignition Residue | 0.02% max |
Assay Percent Range | 99+% |
Packaging | Plastic bottle |
Linear Formula | HO2CCH2C(OH)(CO2H)CH2CO2H·H2O |
Quantity | 2.5 kg |
Merck Index | 15,2325 |
Solubility Information | Solubility in water: 676g/L (25°C). Other solubilities: 2.17g/100g ether - 0.007g/100g chloroform, 15.43g/100g amyl alcohol - 5.98g/100g amyl acetate, 5.28g/100g ethyl acetate - 197g/100g methanol, 62.8g/100g propanol |
Formula Weight | 210.15 |
Show More |
Safety and Handling
Product Identifier
- Citric acid monohydrate
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'