missing translation for 'onlineSavingsMsg'
Learn More

Citric acid, disodium salt sesquihydrate, 99%, pure, Thermo Scientific Chemicals

Catalog Number 10580014
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
1545832
C12H18Na4O17
526.22
MFCD00150445
HGPVLOQNBSHYEI-UHFFFAOYNA-J
unii-cz1032cekr, cz1032cekr, sodium citrate dibasic sesquihydrate, disodium citrate sesquihydrate, disodium hydrogen citrate 1.5 h2o, sodium hydrogen citrate sesquihydrate, citric acid, disodium salt, sesquihydrate, tetrasodium, 3-carboxy-3-hydroxypentanedioate, trihydrate, sodium citrate dibasic sesquihydrate, purum p.a t, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, disodium salt, hydrate 2:3
117063816
tetrasodium;2-(carboxymethyl)-2-hydroxybutanedioate;trihydrate
C(C(=O)O)C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)O)C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.[Na+].[Na+].[Na+].[Na+]
This item is not returnable. View return policy
1545832
C12H18Na4O17
526.22
MFCD00150445
HGPVLOQNBSHYEI-UHFFFAOYNA-J
unii-cz1032cekr, cz1032cekr, sodium citrate dibasic sesquihydrate, disodium citrate sesquihydrate, disodium hydrogen citrate 1.5 h2o, sodium hydrogen citrate sesquihydrate, citric acid, disodium salt, sesquihydrate, tetrasodium, 3-carboxy-3-hydroxypentanedioate, trihydrate, sodium citrate dibasic sesquihydrate, purum p.a t, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, disodium salt, hydrate 2:3
117063816
tetrasodium;2-(carboxymethyl)-2-hydroxybutanedioate;trihydrate
C(C(=O)O)C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)O)C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.[Na+].[Na+].[Na+].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 149.0°C
Color White
pH 4.9 to 5.2 (3% soln.)
Infrared Spectrum Authentic
Assay Percent Range 0.99
Packaging Plastic bottle
Linear Formula 1·5H2O
Quantity 250 g
Merck Index 14, 2326
Solubility Information Solubility in water: 500g/L (20°C).
Formula Weight 263.1
Percent Purity 0.99
Grade Pure
Physical Form Powder
Chemical Name or Material Citric acid, disodium salt sesquihydrate, Pure
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'