missing translation for 'onlineSavingsMsg'
Learn More

Citric acid diammonium salt, 99+%, Thermo Scientific Chemicals

Catalog Number p-7050004
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy
3012-65-5
C6H14N2O7
226.19
MFCD00013068
YXVFQADLFFNVDS-UHFFFAOYSA-N
ammonium citrate dibasic, ammonium hydrogencitrate, diammonium hydrogen citrate, citric acid diammonium salt, citric acid ammonium salt, di-ammonium hydrogen citrate, citro diamine, citric acid diamine, ammonium citrate, dibasic, acs, ammonium citrate dibasic, puriss
13710713
CHEBI:63076
azane;2-hydroxypropane-1,2,3-tricarboxylic acid
[NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O
This item is not returnable. View return policy
3012-65-5
C6H14N2O7
226.19
MFCD00013068
YXVFQADLFFNVDS-UHFFFAOYSA-N
ammonium citrate dibasic, ammonium hydrogencitrate, diammonium hydrogen citrate, citric acid diammonium salt, citric acid ammonium salt, di-ammonium hydrogen citrate, citro diamine, citric acid diamine, ammonium citrate, dibasic, acs, ammonium citrate dibasic, puriss
13710713
CHEBI:63076
azane;2-hydroxypropane-1,2,3-tricarboxylic acid
[NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O

Citric acid diammonium salt is used in pharmaceuticals, rust proofing, cotton printing and plasticizers. It is used as a reagent for the determination of phosphate in fertilizers, and as a buffering agent, chelating agent and source of nitrogen in chicken food supplements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Density 1.48 g/mL
Odor Odorless
Linear Formula HOC(CO2H)(CH2CO2NH4)2
Quantity 100 g
Merck Index 14512
Solubility Information Soluble in water. Slightiy soluble in alcohol.
Formula Weight 226.19
Percent Purity ≥99%
Physical Form Granules
Chemical Name or Material Citric acid diammonium salt
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'