missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Citrate, 0.5M buffer soln., pH 5.5

Catalog Number p-7058188
Click to view available options
Quantity:
250 mL
500 mL
This item is not returnable. View return policy
6132-04-3
This item is not returnable. View return policy
6132-04-3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Concentration 0.5M buffer soln.
Molecular Weight (g/mol) 294.10
pH 5.5
Quantity 250 mL
Odor Odorless
Content And Storage Keep cold
Form Liquid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'