missing translation for 'onlineSavingsMsg'
Learn More

Citraconic anhydride, 98%, Acros Organics

£11.60 - £206.00

Chemical Identifiers

CAS 616-02-4
Molecular Formula C5H4O3
Molecular Weight (g/mol) 112.08
MDL Number MFCD00005522
InChI Key AYKYXWQEBUNJCN-UHFFFAOYSA-N
Synonym citraconic anhydride, 2,5-furandione, 3-methyl, 2-methylmaleic anhydride, methylmaleic anhydride, 3-methylmaleic anhydride, citraconic acid anhydride, monomethylmaleic anhydride, maleic anhydride, methyl, 2-methylmaleicanhydride, unii-105np7pmxx
PubChem CID 12012
IUPAC Name 3-methylfuran-2,5-dione
SMILES CC1=CC(=O)OC1=O
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10006303 SDS View Documents Acros Organics
154790250
25g Glass bottle £11.60
25g View alternative products
Save Up to 
»
EU Stock 1 Please to purchase this item. Need a web account? Register with us today!
10685202 SDS View Documents Acros Organics
154791000
100g Glass bottle £32.20
100g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 04-02-2020
Please to purchase this item. Need a web account? Register with us today!
10524091 SDS View Documents Acros Organics
154792500
250g Glass bottle £62.90
250g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 04-02-2020
Please to purchase this item. Need a web account? Register with us today!
10014103 SDS View Documents Acros Organics
154790010
1kg Glass bottle £206.00
1kg View alternative products
Save Up to 
»
EU Stock 5 Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

616-02-4
112.08
AYKYXWQEBUNJCN-UHFFFAOYSA-N
12012
CC1=CC(=O)OC1=O
C5H4O3
MFCD00005522
citraconic anhydride, 2,5-furandione, 3-methyl, 2-methylmaleic anhydride, methylmaleic anhydride, 3-methylmaleic anhydride, citraconic acid anhydride, monomethylmaleic anhydride, maleic anhydride, methyl, 2-methylmaleicanhydride, unii-105np7pmxx
3-methylfuran-2,5-dione

Specifications

Citraconic anhydride
101°C
Glass bottle
1.245
Colorless to Yellow
25g
616-02-4
C5H4O3
17, 440
Solubility in water: decomposes in water. Other solubilities: soluble in ethanol and ether
CC1=CC(=O)OC1=O
112.08
112.08
98%
1.2450g/mL
Authentic
1.4700 to 1.472
213°C
6°C to 7°C
98%
98%
MFCD00005522
citraconic anhydride, 2,5-furandione, 3-methyl, 2-methylmaleic anhydride, methylmaleic anhydride, 3-methylmaleic anhydride, citraconic acid anhydride, monomethylmaleic anhydride, maleic anhydride, methyl, 2-methylmaleicanhydride, unii-105np7pmxx
AYKYXWQEBUNJCN-UHFFFAOYSA-N
3-methylfuran-2,5-dione
12012
Liquid
Safety and Handling

Safety and Handling

Product Identifier
  • Citraconic anhydride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .