Learn More
Tocris Bioscience™ CITCO
Selective CAR agonist
Brand:  Tocris Bioscience™ 368310
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13463414
  • £132.30 / 10mg
Estimated Shipment
04-04-2019
Add to basket
Description
Description
Constitutive androstane receptor (CAR) agonist (EC 50 = 49 nM). Displays > 100-fold selectivity over PXR receptors and no activity at LXR, ERα, ERβ, PPAR, RAR, FXR, VDR and THR. Induces CAR nuclear translocation and expression of CYP2B6 in hepatocytes in vitro.
Specifications
Specifications
CITCO
>98
10mg
CITCO
436.74