missing translation for 'onlineSavingsMsg'
Learn More

Citalopram hydrobromide, Tocris Bioscience™

Highly potent and selective 5-HT uptake inhibitor

Brand:  Tocris (Bio-Techne) 142750

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13459483

  • £461.52 / 50mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Highly selective and potent 5-HT uptake inhibitor with no effect on noradrenalin or dopamine uptake (IC50 values are 1.8, 8800 and 41000 nM respectively). Has negligible activity at a wide range of receptors.
Specifications

Chemical Identifiers

59729-32-7
405.311
citalopram hydrobromide, citalopram hbr, cipramil, celexa, 1-3-dimethylamino propyl-1-4-fluorophenyl-1,3-dihydroisobenzofuran-5-carbonitrile hydrobromide, elopram, seropram, citalopram hydrobromide, 1-3-dimethylamino propyl-1-4-fluorophenyl-1,3-dihydro-5-isobenzofurancarbonitrile hydrobromide, citalopram hydrobromide usan
1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile;hydrobromide
C20H22BrFN2O
WIHMBLDNRMIGDW-UHFFFAOYSA-N
77995
CN(C)CCCC1(C2=C(CO1)C=C(C=C2)C#N)C3=CC=C(C=C3)F.Br

Specifications

Citalopram hydrobromide
59729-32-7
citalopram hydrobromide, citalopram hbr, cipramil, celexa, 1-3-dimethylamino propyl-1-4-fluorophenyl-1,3-dihydroisobenzofuran-5-carbonitrile hydrobromide, elopram, seropram, citalopram hydrobromide, 1-3-dimethylamino propyl-1-4-fluorophenyl-1,3-dihydro-5-isobenzofurancarbonitrile hydrobromide, citalopram hydrobromide usan
CN(C)CCCC1(C2=C(CO1)C=C(C=C2)C#N)C3=CC=C(C=C3)F.Br
405.311
405.31
50mg
C20H22BrFN2O
WIHMBLDNRMIGDW-UHFFFAOYSA-N
1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile;hydrobromide
77995
>99%
Videos