missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Citadel 2000 Tissue Processor

The Epredia™ Citadel 2000 Tissue Processor is a low cost, reliable, time-tested carousel-type tissue processor with convenient touch key operation and a 110-cassette capacity.

Brand:  Epredia™ 69800004

Code : 50

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 12612613

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

The compact design of the Citadel 2000 carousel tissue processor, coupled with its proven processing quality, ease of operation and excellent chemical resistance explains why it has become the tried and trusted choice for thousands of laboratories worldwide.

Features:

  • Specimens are automatically raised from their reagent containers every 10 minutes and then reimmersed
  • Optional vacuum unit for efficient and rapid wax infiltration
  • Unique touch key pad incorporates all processing functions
  • Up to four complete schedules can be programmed
  • Programmable delayed start time
  • Loading and unloading of specimens is made easy with the specially designed tissue basket hanger
  • Random baskets holding 70 cassettes or 110 cassette organiser baskets
  • Smooth surfaces and contoured design facilitate easy cleaning
  • Integral fume hood greatly reduces release of vapours
  • Easy change of reagent containers

Physical Specifications:

  • Electrical Requirements: 100-240 V, 50/60 Hz
  • Diameter: 68 cm (27 in.)
  • Height: 62 cm (25 in.)
  • Weight: 106 lb. (48 kg)

Includes:

  • Basic instrument
  • Instruction manual
  • Ten reagent vessels and two paraffin baths
Specifications

Specifications

Tissue Processor
2 L
220/240 VAC
110 (Organised); 70 (Random)
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers