missing translation for 'onlineSavingsMsg'
Learn More

Cisapride, Tocris Bioscience™

5-HT4 agonist; stimulates intestinal ACh release

Brand:  Tocris (Bio-Techne) 169510

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13459933

  • £120.15 / 10mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

5-HT4 receptor agonist and gastrokinetic agent. Stimulates intestinal acetylcholine release, possibly via 5-HT4 receptor-dependent and -independent mechanisms, leading to increased intestinal motility.
Specifications

Chemical Identifiers

81098-60-4
465.95
cisapride, acenalin, propulsin, alimix, coordinax, +/--cisapride, 4-amino-5-chloro-n-3r,4r-1-3-4-fluorophenoxy propyl-3-methoxypiperidin-4-yl-2-methoxybenzamide, cis-2-methoxy-4-amino-5-chloro-n-1-3-4-fluorophenoxy propyl-3-methoxy-4-piperidinyl benzamide, rel-4-amino-5-chloro-n-1-3r,4s-3-4-fluorophenoxy propyl-3-methoxy-4-piperidinyl-2-methoxybenzamide
4-amino-5-chloro-N-[(3R,4R)-1-[3-(4-fluorophenoxy)propyl]-3-methoxypiperidin-4-yl]-2-methoxybenzamide
C23H29ClFN3O4
DCSUBABJRXZOMT-IFMALSPDSA-N
13677779
COC1CN(CCC1NC(=O)C2=CC(=C(C=C2OC)N)Cl)CCCOC3=CC=C(C=C3)F

Specifications

Cisapride
81098-60-4
cisapride, acenalin, propulsin, alimix, coordinax, +/--cisapride, 4-amino-5-chloro-n-3r,4r-1-3-4-fluorophenoxy propyl-3-methoxypiperidin-4-yl-2-methoxybenzamide, cis-2-methoxy-4-amino-5-chloro-n-1-3-4-fluorophenoxy propyl-3-methoxy-4-piperidinyl benzamide, rel-4-amino-5-chloro-n-1-3r,4s-3-4-fluorophenoxy propyl-3-methoxy-4-piperidinyl-2-methoxybenzamide
COC1CN(CCC1NC(=O)C2=CC(=C(C=C2OC)N)Cl)CCCOC3=CC=C(C=C3)F
465.95
465.95
10mg
C23H29ClFN3O4
DCSUBABJRXZOMT-IFMALSPDSA-N
4-amino-5-chloro-N-[(3R,4R)-1-[3-(4-fluorophenoxy)propyl]-3-methoxypiperidin-4-yl]-2-methoxybenzamide
13677779
>99%
Videos