missing translation for 'onlineSavingsMsg'
Learn More

cis-Dicyclohexano-18-crown-6, 98%, mixture of syn-cis and anti-cis isomers, ACROS Organics™

£39.10 - £124.00

Chemical Identifiers

CAS 16069-36-6
Molecular Formula C20H36O6
Molecular Weight (g/mol) 372.5
MDL Number MFCD00005099
InChI Key BBGKDYHZQOSNMU-UHFFFAOYSA-N
Synonym dicyclohexano-18-crown-6, dicyclohexyl-18-crown-6, cis-dicyclohexano-18-crown-6, dicyclohexo-18-crown-6, perhydrodibenzo-18-crown-6, dch-18-crown-6, icosahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine, dicyclohexyl-18-crown-6 ether, dicyclohexyl 18-crown-6, ccris 3612
PubChem CID 85955
SMILES C1CCC2C(C1)OCCOCCOC3CCCCC3OCCOCCO2
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10420721
SDS View Documents
Acros Organics
277520025
2.5g Glass bottle
£39.10
2.50g
View alternative products
Save Up to 
»
In stock, ready to ship: 1
Please to purchase this item. Need a web account? Register with us today!
10043500
SDS View Documents
Acros Organics
277520100
10g Glass bottle
£124.00
10g
View alternative products
Save Up to 
»
In stock, ready to ship: 3
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Chelation/Complexation reagent
Specifications

Chemical Identifiers

16069-36-6
372.5
BBGKDYHZQOSNMU-UHFFFAOYSA-N
85955
C20H36O6
MFCD00005099
dicyclohexano-18-crown-6, dicyclohexyl-18-crown-6, cis-dicyclohexano-18-crown-6, dicyclohexo-18-crown-6, perhydrodibenzo-18-crown-6, dch-18-crown-6, icosahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine, dicyclohexyl-18-crown-6 ether, dicyclohexyl 18-crown-6, ccris 3612
C1CCC2C(C1)OCCOCCOC3CCCCC3OCCOCCO2

Specifications

cis-Dicyclohexano-18-crown-6
Glass bottle
White to Beige
2.5g
16069-36-6
C20H36O6
04,142; 05,207; 06,174; 09,126; 12,147; 01,1046; 03,261; 04,442; 06,529; 12,441; 16,307
Solubility in water: slightly soluble. Other solubilities: soluble in chloroform and acetone
C1CCC2C(C1)OCCOCCOC3CCCCC3OCCOCCO2
85955
Crystalline Solid
Authentic
181°C to 183°C
47°C to 53°C
98%
97.5% min. (GC)
MFCD00005099
dicyclohexano-18-crown-6, dicyclohexyl-18-crown-6, cis-dicyclohexano-18-crown-6, dicyclohexo-18-crown-6, perhydrodibenzo-18-crown-6, dch-18-crown-6, icosahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine, dicyclohexyl-18-crown-6 ether, dicyclohexyl 18-crown-6, ccris 3612
BBGKDYHZQOSNMU-UHFFFAOYSA-N
372.5
372.5
98%
SDS
Product Certifications