missing translation for 'onlineSavingsMsg'
Learn More

cis-Dicyclohexano-18-crown-6, 98%, mixture of syn-cis and anti-cis isomers, Thermo Scientific Chemicals

Catalog Number 10043500
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
16069-36-6
C20H36O6
372.5
MFCD00005099
BBGKDYHZQOSNMU-UHFFFAOYSA-N
dicyclohexano-18-crown-6, dicyclohexyl-18-crown-6, cis-dicyclohexano-18-crown-6, dicyclohexo-18-crown-6, perhydrodibenzo-18-crown-6, dch-18-crown-6, icosahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine, dicyclohexyl-18-crown-6 ether, dicyclohexyl 18-crown-6, ccris 3612
85955
C1CCC2C(C1)OCCOCCOC3CCCCC3OCCOCCO2
This item is not returnable. View return policy
16069-36-6
C20H36O6
372.5
MFCD00005099
BBGKDYHZQOSNMU-UHFFFAOYSA-N
dicyclohexano-18-crown-6, dicyclohexyl-18-crown-6, cis-dicyclohexano-18-crown-6, dicyclohexo-18-crown-6, perhydrodibenzo-18-crown-6, dch-18-crown-6, icosahydrodibenzo b,k 1,4,7,10,13,16 hexaoxacyclooctadecine, dicyclohexyl-18-crown-6 ether, dicyclohexyl 18-crown-6, ccris 3612
85955
C1CCC2C(C1)OCCOCCOC3CCCCC3OCCOCCO2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY
Melting Point 47°C to 53°C
Color White to Beige
Boiling Point 181°C to 183°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 10 g
Fieser 04,142; 05,207; 06,174; 09,126; 12,147; 01,1046; 03,261; 04,442; 06,529; 12,441; 16,307
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in chloroform and acetone
Formula Weight 372.5
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material cis-Dicyclohexano-18-crown-6, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'