missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ cis-Dichlorobis(ethylenediamine)platinum(II), 99.9% (metals basis)
Brand: Alfa Aesar™ 039667.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
cis-Dichlorobis(ethylenediamine)platinum(II) | |
386.19 | |
MFCD00009881 |
Specifications
cis-Dichlorobis(ethylenediamine)platinum(II) | |
99.9% | |
386.19 | |
1g |
Crystalline | |
(metals basis) | |
MFCD00009881 | |
Hygroscopic |