missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ cis-Dichlorobis(ethylenediamine)platinum(II), 99.9% (metals basis)

Brand:  Alfa Aesar™ 039667.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11364617

  • £549.19 / 5g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

cis-Dichlorobis(ethylenediamine)platinum(II)
386.19
MFCD00009881

Specifications

cis-Dichlorobis(ethylenediamine)platinum(II)
99.9%
386.19
5g
Crystalline
(metals basis)
MFCD00009881
Hygroscopic
Videos
SDS