missing translation for 'onlineSavingsMsg'
Learn More

cis-ACPD, Tocris Bioscience™

Potent NMDA agonist. Also group II mGluR agonist

Brand:  Tocris (Bio-Techne) 018650

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13447463

  • £482.16 / 50mg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

1:1 mixture of (1R,3R)- and (1S,3S)-ACPD. Potent NMDA receptor agonist and selective group II mGluR agonist (EC50 values are 13, 50, >300 and >300 μM at mGluR2, mGluR4, mGluR1 and mGluR5 respectively). Exhibits proconvulsant activity in vivo.

(±)-trans-ACPD and (1S,3R)-ACPD also available.

Specifications

Chemical Identifiers

477331-06-9
173.168
1s,3s-1-aminocyclopentane-1,3-dicarboxylic acid, chembl29726, trans-acpd, cis-acpd, +/--trans-acpd, 1s, 3r-acpd, 1-amino-cis-cyclopentane-1,3-dicarboxylic acid, tocris-0186, tocris-0187, tocris-0284
(1S,3S)-1-aminocyclopentane-1,3-dicarboxylic acid
C7H11NO4
YFYNOWXBIBKGHB-FFWSUHOLSA-N
6604704
C1CC(CC1C(=O)O)(C(=O)O)N

Specifications

50mg
477331-06-9
1s,3s-1-aminocyclopentane-1,3-dicarboxylic acid, chembl29726, trans-acpd, cis-acpd, +/--trans-acpd, 1s, 3r-acpd, 1-amino-cis-cyclopentane-1,3-dicarboxylic acid, tocris-0186, tocris-0187, tocris-0284
C1CC(CC1C(=O)O)(C(=O)O)N
173.168
173.17
cis-ACPD
C7H11NO4
YFYNOWXBIBKGHB-FFWSUHOLSA-N
(1S,3S)-1-aminocyclopentane-1,3-dicarboxylic acid
6604704
>98%
Videos
Documents