missing translation for 'onlineSavingsMsg'
Learn More

cis-Aconitic acid, tech. 85%, Thermo Scientific Chemicals

Catalog Number p-7022994
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
585-84-2
C6H6O6
174.11
MFCD00063184
GTZCVFVGUGFEME-HNQUOIGGSA-N
cis-aconitic acid, cis-aconitate, achilleic acid, citridinic acid, equisetic acid, pyrocitric acid, achilleaic acid, citridic acid, cis-aconic acid, aconitic acid
643757
CHEBI:32805
OC(=O)C\C(=C/C(O)=O)C(O)=O
This item is not returnable. View return policy
585-84-2
C6H6O6
174.11
MFCD00063184
GTZCVFVGUGFEME-HNQUOIGGSA-N
cis-aconitic acid, cis-aconitate, achilleic acid, citridinic acid, equisetic acid, pyrocitric acid, achilleaic acid, citridic acid, cis-aconic acid, aconitic acid
643757
CHEBI:32805
OC(=O)C\C(=C/C(O)=O)C(O)=O

cis-Aconitic acid is used as an intermediate in the production of substances pharmaceuticals. It is widely used in chemical research and as reagent. It acts as a cis-isomer of aconitic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
cis-Aconitic acid is used as an intermediate in the production of substances pharmaceuticals. It is widely used in chemical research and as reagent. It acts as a cis-isomer of aconitic acid.

Solubility
Soluble in water, alcohol. Slightly soluble in ether.
TRUSTED_SUSTAINABILITY
Melting Point 115°C to 120°C
Assay Percent Range tech. 90%
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Soluble in water,alcohol. Slightly soluble in ether.
IUPAC Name (Z)-prop-1-ene-1,2,3-tricarboxylic acid
Formula Weight 174.11
Percent Purity 90%
Grade Technical
Chemical Name or Material cis-Aconitic Acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'