missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals cis-4-Hydroxy-D-proline, 99%

Catalog Number p-3757503
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
2584-71-6
C5H9NO3
131.13
MFCD00005252
PMMYEEVYMWASQN-QWWZWVQMSA-N
cis-4-hydroxy-d-proline, h-d-cis-hyp-oh, 2r,4r-4-hydroxypyrrolidine-2-carboxylic acid, d-cis-hydroxyproline, d-allo-hydroxyproline, d-proline, 4-hydroxy-, 4r, 4r-4-hydroxy-d-proline, d-cis-4-hydroxyproline, cis-4-hydroxy-d-prolin, 2r,4r-4-hydroxy-pyrrolidine-2-carboxylic acid
440014
CHEBI:16231
(2R,4R)-4-hydroxypyrrolidine-2-carboxylic acid
O[C@H]1CN[C@H](C1)C(O)=O
This item is not returnable. View return policy
2584-71-6
C5H9NO3
131.13
MFCD00005252
PMMYEEVYMWASQN-QWWZWVQMSA-N
cis-4-hydroxy-d-proline, h-d-cis-hyp-oh, 2r,4r-4-hydroxypyrrolidine-2-carboxylic acid, d-cis-hydroxyproline, d-allo-hydroxyproline, d-proline, 4-hydroxy-, 4r, 4r-4-hydroxy-d-proline, d-cis-4-hydroxyproline, cis-4-hydroxy-d-prolin, 2r,4r-4-hydroxy-pyrrolidine-2-carboxylic acid
440014
CHEBI:16231
(2R,4R)-4-hydroxypyrrolidine-2-carboxylic acid
O[C@H]1CN[C@H](C1)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Assay Percent Range 99%
Infrared Spectrum Authentic
Beilstein 22, I, 546
Merck Index 10, 4762
Packaging Glass Bottle
Specific Rotation +56° to +60° (24°C, 589nm) (c=2, water)
Formula Weight 131.13
Percent Purity ≥98.5%
Specific Rotation Condition +58° (24°C c=2,water)
Quantity 250 mg
Physical Form Powder
Chemical Name or Material cis-4-Hydroxy-D-proline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'