missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-4-Amino-N-Boc-L-proline methyl ester hydrochloride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
cis-4-Amino-N-Boc-L-proline methyl ester hydrochloride is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
cis-4-Amino-N-Boc-L-proline methyl ester hydrochloride is used as pharmaceutical intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
cis-4-Amino-N-Boc-L-proline methyl ester hydrochloride is used as pharmaceutical intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Melting Point | ∽185°C (decomposition) |
Quantity | 1 g |
Optical Rotation | −40° (c=2 in Methanol) |
IUPAC Name | 1-O-tert-butyl 2-O-methyl (2S,4S)-4-aminopyrrolidine-1,2-dicarboxylate;hydrochloride |
Formula Weight | 280.75 |
Percent Purity | 97% |
Chemical Name or Material | cis-4-Amino-N-Boc-L-proline methyl ester hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'