missing translation for 'onlineSavingsMsg'
Learn More

cis-4-Amino-1-cyclohexanecarboxylic acid, 97%, ACROS Organics™

Brand:  Acros Organics 368330010

Code : 29

Additional Details : CAS Number : 3685-23-2 Weight : 0.05100kg


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10406452

  • £41.70 / 1g


EU Stock 2
Add to basket


Specifications

Chemical Identifiers

cis-4-Amino-1-cyclohexanecarboxylic acid
3685-23-2
97%
143.19
97%
≥96.0%
C7H13NO2
MFCD00191730

Specifications

cis-4-Amino-1-cyclohexanecarboxylic acid
Glass bottle
Authentic
Off-white
Powder
97%
143.19
14, IV, 981
97%
3685-23-2
278° to 283°C
1g
≥96.0%
C7H13NO2
MFCD00191730
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • cis-4-Amino-1-cyclohexanecarboxylic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS