missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ cis-2-(Boc-amino)cyclopropanemethanol, 97%

Brand:  Alfa Aesar™ H62795.03

Code : NEW

Additional Details : CAS Number : 170299-61-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15449268

  • £835.90 / 1g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Description

Description

Applications
cis-2-(Boc-amino)cyclopropanemethanol is employed as a intermediate for chemical research.

Notes
Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

cis-2-(Boc-amino)cyclopropanemethanol
C9H17NO3
MFCD03844611
170299-61-3
187.23
tert-Butyl cis-(2-hydroxymethyl)cyclopropylcarbamate; cis-1-(Boc-amino)-2-(hydroxymethyl)cyclopropane

Specifications

cis-2-(Boc-amino)cyclopropanemethanol
97%
187.23
tert-Butyl cis-(2-hydroxymethyl)cyclopropylcarbamate; cis-1-(Boc-amino)-2-(hydroxymethyl)cyclopropane
170299-61-3
C9H17NO3
MFCD03844611
1g
Videos
SDS