missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ cis-2-(Boc-amino)cyclohexanecarboxylic acid, 97%

Brand:  Alfa Aesar™ H62600.MD

Code : NEW

Additional Details : CAS Number : 63216-49-9 Weight : 0.00025kg

Quantity 250mg

 View more versions of this product

Product Code. 15427626

  • £19.50 / 250mg


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

cis-2-(Boc-amino)cyclohexanecarboxylic acid
243.29
cis-2-tert-Butoxycarbonylaminocyclohexanecarboxylic acid; Boc-1,2-cis-ACHC-OH
63216-49-9
MFCD01863244

Specifications

cis-2-(Boc-amino)cyclohexanecarboxylic acid
Solid
243.29
127° to 131°C
250mg
63216-49-9
97%
MFCD01863244
cis-2-tert-Butoxycarbonylaminocyclohexanecarboxylic acid; Boc-1,2-cis-ACHC-OH
Videos
SDS