missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ cis-2-(Benzyloxycarbonylamino)cyclohexanecarboxylic acid, 97%

Brand:  Alfa Aesar™ H62548.03

Code : NEW

Additional Details : CAS Number : 54867-08-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15437556

  • £56.60 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

cis-2-(Benzyloxycarbonylamino)cyclohexanecarboxylic acid
277.31
Z-1,2-cis-ACHC-OH; Z-cis-2-aminocyclohexanecarboxylic acid
54867-08-2
MFCD01863245

Specifications

cis-2-(Benzyloxycarbonylamino)cyclohexanecarboxylic acid
Solid
277.31
Z-1,2-cis-ACHC-OH; Z-cis-2-aminocyclohexanecarboxylic acid
54867-08-2
97%
MFCD01863245
1g
Videos
SDS