missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Amino-1-cyclohexanecarboxamide, 98+%, Thermo Scientific Chemicals

Catalog Number 10482825
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
115014-77-2
C7H14N2O
142.2
MFCD00143979
DXNKJRKPVQVDJW-WDSKDSINSA-N
cyclohexanecarboxamide,2-amino-, 1r,2s-rel, 1s-2beta-aminocyclohexane-1alpha-carboxamide
30069041
(1S,2S)-2-aminocyclohexane-1-carboxamide
C1CCC(C(C1)C(=O)N)N
This item is not returnable. View return policy
115014-77-2
C7H14N2O
142.2
MFCD00143979
DXNKJRKPVQVDJW-WDSKDSINSA-N
cyclohexanecarboxamide,2-amino-, 1r,2s-rel, 1s-2beta-aminocyclohexane-1alpha-carboxamide
30069041
(1S,2S)-2-aminocyclohexane-1-carboxamide
C1CCC(C(C1)C(=O)N)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 124°C to 127°C
Color White
Assay Percent Range 98+%
Infrared Spectrum Authentic
Packaging Glass Bottle
Solubility Information Solubility in water: .. Other solubilities: soluble in most common organic solvents
Formula Weight 142.2
Percent Purity ≥98%
Quantity 1 g
Physical Form Crystalline Powder
Chemical Name or Material cis-2-Amino-1-cyclohexanecarboxamide, 98+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • cis-2-Amino-1-cyclohexanecarboxamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'