missing translation for 'onlineSavingsMsg'
Learn More

cis-2,6-Dimethylpiperidine, 97+%, Thermo Scientific™

Catalog Number p-119590
Click to view available options
Quantity:
5 g
100 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
766-17-6
C7H15N
113.2
MFCD00005986
SDGKUVSVPIIUCF-KNVOCYPGSA-N
cis-2,6-dimethylpiperidine, 2r,6s-2,6-dimethylpiperidine, cis-hexahydro-2,6-lutidine, cis-2,6-lupetidine, piperidin, 2e,6e-dimethyl, cis-2,6-dimethyl-piperidine, cis-lupetidine, cis-nanophine, prestwick0_000666, prestwick1_000666
641768
(2R,6S)-2,6-dimethylpiperidine
CC1CCCC(N1)C
This item is not returnable. View return policy
766-17-6
C7H15N
113.2
MFCD00005986
SDGKUVSVPIIUCF-KNVOCYPGSA-N
cis-2,6-dimethylpiperidine, 2r,6s-2,6-dimethylpiperidine, cis-hexahydro-2,6-lutidine, cis-2,6-lupetidine, piperidin, 2e,6e-dimethyl, cis-2,6-dimethyl-piperidine, cis-lupetidine, cis-nanophine, prestwick0_000666, prestwick1_000666
641768
(2R,6S)-2,6-dimethylpiperidine
CC1CCCC(N1)C

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 97.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8400g/mL
Boiling Point 127°C to 128°C
Flash Point 11°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Refractive Index 1.4384 to 1.4404
Quantity 5 g
UN Number 2733
Beilstein 20, 108
Fieser 02,156
Specific Gravity 0.84
Solubility Information Solubility in water: miscible. Other solubilities: soluble in most organic solvents
Formula Weight 113.2
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material cis-2, 6-Dimethylpiperidine, 99%, min. 95% cis-isomer
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • cis-2,6-Dimethylpiperidine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'